Skip to Content
Merck
All Photos(1)

Key Documents

665916

Sigma-Aldrich

Yttrium(III) tris(isopropoxide)

Synonym(s):

2-Propanol yttrium(III) salt, Isopropyl alcohol yttrium(III) salt, Tris(isopropoxy) yttrium(III)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21O3Y
CAS Number:
Molecular Weight:
266.17
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: yttrium
reagent type: catalyst

SMILES string

CC(C)O[Y](OC(C)C)OC(C)C

InChI

1S/3C3H7O.Y/c3*1-3(2)4;/h3*3H,1-2H3;/q3*-1;+3

InChI key

PYLIDHFYDYRZSC-UHFFFAOYSA-N

Application

Catalyst for:
  • Stereoselective conjugate addition reactions
  • Ring-opening polymerizations
  • Generation of reactive enolates by enantioselective protonation reactions
  • Cyano-phosphorylation of enone and allylic substitution
  • Synthesis of nanocomposites

Reactant for the preparation of flat HfO2 films
In many asymmetric catalysis applications, glove box and Schlenk techniques should be employed to prevent exposure of the rare earth catalyst to air and moisture, which can be detrimental to the reaction outcome. Solutions of the catalyst should be made using anhydrous solvents and used shortly after preparation.

Legal Information

Product of Kojundo Chemical Laboratory Co., Ltd

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Related Content

Professor Shibasaki's research focuses on the development of novel cooperative asymmetric catalytic systems that allowed for streamlined synthesis of enantioenriched high-value chiral building blocks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service