Skip to Content
Merck
All Photos(1)

Key Documents

608130

Sigma-Aldrich

L-Serine-13C3,15N

98 atom % 15N, 98 atom % 13C, 95% (CP)

Synonym(s):

L-Serine-UL-13C,15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO13CH213CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
109.06
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 13C
98 atom % 15N

Assay

95% (CP)

form

solid

optical activity

[α]25/D +14.6°, c = 2 in 1 M HCl

technique(s)

bio NMR: suitable

mp

222 °C (dec.) (lit.)

mass shift

M+4

SMILES string

[15NH2][13C@@H]([13CH2]O)[13C](O)=O

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)/t2-/m0/s1/i1+1,2+1,3+1,4+1

InChI key

MTCFGRXMJLQNBG-UVYXLFMMSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc Hennequart et al.
Nature communications, 12(1), 6176-6176 (2021-10-28)
Serine is a non-essential amino acid that is critical for tumour proliferation and depletion of circulating serine results in reduced tumour growth and increased survival in various cancer models. While many cancer cells cultured in a standard tissue culture medium
Dominic G Roy et al.
Cell metabolism, 31(2), 250-266 (2020-02-06)
Epigenetic modifications on DNA and histones regulate gene expression by modulating chromatin accessibility to transcription machinery. Here we identify methionine as a key nutrient affecting epigenetic reprogramming in CD4+ T helper (Th) cells. Using metabolomics, we showed that methionine is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service