Skip to Content
Merck
All Photos(1)

Key Documents

516767

Sigma-Aldrich

5-Bromo-2-chloroanisole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(Cl)OCH3
CAS Number:
Molecular Weight:
221.48
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.587 (lit.)

bp

240-245 °C/748 mmHg (lit.)

mp

27-28 °C (lit.)

density

1.631 g/mL at 25 °C (lit.)

SMILES string

COc1cc(Br)ccc1Cl

InChI

1S/C7H6BrClO/c1-10-7-4-5(8)2-3-6(7)9/h2-4H,1H3

InChI key

UAMVKOTWSHJOSY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J I Cacho et al.
Journal of chromatography. A, 1475, 74-79 (2016-11-15)
A solventless analytical method is proposed for analyzing the compounds responsible for cork taint in cork stoppers. Direct sample introduction (DSI) is evaluated as a sample introduction system for the gas chromatography-mass spectrometry (GC-MS) determination of four haloanisoles (HAs) in
Novel Stereoselective Syntheses of the Fused Benzazepine Dopamine D1 Antagonist (6a S, 13b R)-11-Chloro-6, 6a, 7, 8, 9, 13b-hexahydro-7-methyl-5 H-benzo [d] naphth [2, 1-b] azepin-12-ol (Sch 39166): 1. Aziridinium Salt Based Syntheses.
Draper RW, et al.
Organic Process Research & Development, 2(3), 175-185 (1998)
David S Millan et al.
Journal of medicinal chemistry, 54(22), 7797-7814 (2011-09-06)
This paper describes the identification and optimization of a novel series of DFG-out binding p38 inhibitors as inhaled agents for the treatment of chronic obstructive pulmonary disease. Structure based drug design and "inhalation by design" principles have been applied to
M Pastor-Belda et al.
Journal of chromatography. A, 1509, 43-49 (2017-06-24)
Glyoxal (GO) and methylglyoxal (MGO) are α-oxoaldehydes that can be used as urinary diabetes markers. In this study, their levels were measured using a sample preparation procedure based on salting-out assisted liquid-liquid extraction (SALLE) and dispersive liquid-liquid microextraction (DLLME) combined
Tyrosine Hydroxylase Inhibitors. Synthesis and Activity of Substituted Aromatic Amino Acids.
Saari WS, et al.
Journal of Medicinal Chemistry, 10(6), 1008-1014 (1967)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service