Skip to Content
Merck
All Photos(1)

Key Documents

451851

Sigma-Aldrich

Sodium dodecyl-d25 sulfate

≥98 atom % D, ≥98% (CP)

Synonym(s):

Dodecyl-d25 sulfate sodium salt, SDS-d25, sodium lauryl sulfate-d25

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3(CD2)11OSO3Na
CAS Number:
Molecular Weight:
313.53
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

technique(s)

NMR: suitable
bio NMR: suitable

mp

204-207 °C (lit.)

mass shift

M+25

SMILES string

[Na+].[2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])OS([O-])(=O)=O

InChI

1S/C12H26O4S.Na/c1-2-3-4-5-6-7-8-9-10-11-12-16-17(13,14)15;/h2-12H2,1H3,(H,13,14,15);/q;+1/p-1/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2;

InChI key

DBMJMQXJHONAFJ-YCGROXIYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Purohit et al.
International journal of cosmetic science, 36(4), 379-385 (2014-05-16)
Stratum corneum (SC) lipids are known to play an important role in barrier properties of skin by maintaining the optimal hydration levels. The disruption of SC lipids by cleanser surfactants is believed to lead to dry skin damage which can
Amrita Roy Choudhury et al.
PloS one, 10(8), e0135455-e0135455 (2015-08-21)
We present a 3D model of the four transmembrane (TM) helical regions of bilitranslocase (BTL), a structurally uncharacterized protein that transports organic anions across the cell membrane. The model was computed by considering helix-helix interactions as primary constraints, using Monte
Guanhua Yang et al.
Biopolymers, 104(1), 11-20 (2014-12-05)
Caveolin-1 is a main structural component of caveolae and essential for the invagination of caveolae by forming a hairpin-shaped structure in the membrane-inserting domain (residues, 102-122). In this article, we determined the tertiary structures of the peptides comprising residues 93-126
Lars Erik Uggerhøj et al.
FEBS letters, 588(17), 3291-3297 (2014-07-27)
The inclusion of peptoid monomers into antimicrobial peptides (AMPs) increases their proteolytic resistance, but introduces conformational flexibility (reduced hydrogen bonding ability and cis/trans isomerism). We here use NMR spectroscopy to answer how the insertion of a peptoid monomer influences the
Ludovic Carlier et al.
Biophysical chemistry, 196, 40-52 (2014-10-08)
Plasticins (PTC) are dermaseptin-related antimicrobial peptides characterized by a large number of leucine and glycine residues arranged in GXXXG motifs that are often described to promote helix association within biological membranes. We report the structure and interaction properties of two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service