Skip to Content
Merck
All Photos(1)

Key Documents

450596

Sigma-Aldrich

Poly(ethylene glycol) bis(2-ethylhexanoate)

average Mn ~650

Synonym(s):

PEG bis(2-ethylhexanoate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CO(OCH2CH2)nO2CCH(C2H5)(CH2)3CH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

mol wt

average Mn ~650

refractive index

n20/D 1.453 (lit.)

viscosity

50 cP(25 °C)(lit.)

mp

−48 °C (lit.)

solubility

H2O: insoluble
hexane, toluene, ethanol, and acetone: soluble

density

1.02 g/mL at 25 °C (lit.)

SMILES string

OCCO.CCCCC(CC)C(O)=O

InChI

1S/C18H34O4/c1-5-9-11-15(7-3)17(19)21-13-14-22-18(20)16(8-4)12-10-6-2/h15-16H,5-14H2,1-4H3

InChI key

XCQFTFZCTGZCNP-UHFFFAOYSA-N

General description

Poly(ethylene glycol) bis(2-ethylhexanoate) (PEG-EH) is an ethylene acrylic elastomer that can be used as a low volatile plasticizer.

Application

PEG-EH can form a blend with polyvinylidene fluoride (PVDF) which can be used in the preparation of electrolytes for lithium batteries. It is also used as an additive for the development of ionization methods for mass spectroscopy.

Features and Benefits

Low volatility and good heat resistance. Compatible with a variety of polymers including polyesters and polyamides.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel microporous poly (vinylidene fluoride) blend electrolytes for lithium-ion batteries
Wang H, et al.
Journal of the Electrochemical Society, 147(8), 2853-2861 (2000)
A novel all-solid electrolyte based on a co-polymer of poly-(methoxy/hexadecal-poly (ethylene glycol) methacrylate) for lithium-ion cell
Zuo X, et al.
Journal of Materials Chemistry, 22(41), 22265-22271 (2012)
Jonathan P Williams et al.
Rapid communications in mass spectrometry : RCM, 21(11), 1693-1704 (2007-05-03)
A rapid method for the characterisation of polyglycol esters and ethers is described which uses accurate mass desorption electrospray ionisation (DESI) quadrupole time-of-flight mass spectrometry (Q-ToFMS). The results are combined with newly developed software which aids the interpretation of product
Research and development topics in Analytical Chemistry. Investigation of a cation exchange separation method for the determination of transition metal ions in anaerobic sealants
O'Dea P, et al.
Anal. Proc., 28(3), 82-84 (1991)
Klaus E Andersen et al.
Contact dermatitis, 70(2), 112-116 (2013-10-25)
Allergic reactions to spectacle frames are not unusual. A patient had a reproducible strong allergic patch test reaction to scrapings from the plastic material, and negative patch test results with available spectacle frame chemicals. To identify the culprit allergen in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service