Skip to Content
Merck
All Photos(2)

Key Documents

400076

Sigma-Aldrich

(Ethylenediamine)palladium(II) chloride

97%

Synonym(s):

NSC 153155, NSC 209490, Dichloro(1,2-diaminoethane)palladium, Dichloro(ethylenediamine)palladium(II), Palladium(II) chloride ethylenediamine complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(H2NCH2CH2NH2)Cl2
CAS Number:
Molecular Weight:
237.42
Beilstein:
4934132
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

for synthesis

Assay

97%

form

solid

reaction suitability

core: palladium
reaction type: Cross Couplings
reagent type: catalyst

mp

>278 °C (lit.)

SMILES string

Cl[Pd]Cl.NCCN

InChI

1S/C2H8N2.2ClH.Pd/c3-1-2-4;;;/h1-4H2;2*1H;/q;;;+2/p-2

InChI key

CAYKJANQVKIYPJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Synthesis of 6-(substituted benzyl)imidazo[2,1-b][1,3]thiazole
  • Sonogashira-coupling reactions

Reactant for:
  • Self-assembly of a ring-in-ring complex
  • Metal binding with glycose phosphates
  • Synthesis of mixed-mtal, mixed-pyrimidine self-assembling metallacalix[n]arenes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service