Skip to Content
Merck
All Photos(1)

Key Documents

361127

Sigma-Aldrich

4-Chloro-3-nitrophenol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)OH
CAS Number:
Molecular Weight:
173.55
Beilstein:
2094546
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

125-128 °C (lit.)

SMILES string

Oc1ccc(Cl)c(c1)[N+]([O-])=O

InChI

1S/C6H4ClNO3/c7-5-2-1-4(9)3-6(5)8(10)11/h1-3,9H

InChI key

JUIKCULGDIZNDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloro-3-nitrophenol is a chloronitrophenol, widely used as an important building block for the synthesis of dyes, drugs, explosives and pesticides. Degradation of 4-chloro-3-nitrophenol by Pseudomonas sp. JHN is reported.

Application

4-Chloro-3-nitrophenol may be used as sole carbon and energy supplement for Pseudomonas sp. JHN.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janmejay Pandey et al.
Applied microbiology and biotechnology, 92(3), 597-607 (2011-06-01)
Burkholderia sp. strain SJ98 (DSM 23195) was previously isolated and characterized for degradation and co-metabolic transformation of a number nitroaromatic compounds. In the present study, we evaluated its metabolic activity on chlorinated nitroaromatic compounds (CNACs). Results obtained during this study
Pankaj Kumar Arora et al.
Scientific reports, 4, 4475-4475 (2014-03-29)
A 4-chloro-3-nitrophenol (4C3NP)-mineralizing bacterium, Pseudomonas sp. JHN was isolated from a waste water sample collected from a chemically-contaminated area, India by an enrichment method. Pseudomonas sp. JHN utilized 4C3NP as a sole carbon and energy source and degraded it with
Surendra Vikram et al.
PloS one, 8(12), e84766-e84766 (2014-01-01)
Biodegradation of para-Nitrophenol (PNP) proceeds via two distinct pathways, having 1,2,3-benzenetriol (BT) and hydroquinone (HQ) as their respective terminal aromatic intermediates. Genes involved in these pathways have already been studied in different PNP degrading bacteria. Burkholderia sp. strain SJ98 degrades

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service