Skip to Content
Merck
All Photos(1)

Key Documents

360732

Sigma-Aldrich

Magnesium alloyed with ~2% nickel

fine powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.23

form

fine powder

contains

1.8-2.0% nickel as enhancer of hydride storage capacity

Application

An active reducing agent and hydride storage material.

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Self-heat. 1 - Skin Irrit. 2 - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service