Skip to Content
Merck
All Photos(2)

Key Documents

338532

Sigma-Aldrich

3-Isoquinolinecarbonitrile

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H6N2
CAS Number:
Molecular Weight:
154.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

126-128 °C (lit.)

SMILES string

N#Cc1cc2ccccc2cn1

InChI

1S/C10H6N2/c11-6-10-5-8-3-1-2-4-9(8)7-12-10/h1-5,7H

InChI key

GXXBVBVNGMCFIU-UHFFFAOYSA-N

Application

3-Isoquinolinecarbonitrile was employed as analytical standard to study the thermal degradation of 2-chlorobenzylidenemalononitrile (CS riot control agent) at elevated temperatures.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy A Kluchinsky et al.
Journal of chromatography. A, 952(1-2), 205-213 (2002-06-18)
2-Chlorobenzylidenemalononitrile (CS riot control agent) has been shown to produce a number of thermal degradation products when dispersed at high temperature. We hypothesized that these CS-derived compounds are formed by energy input from heating during the dispersion process. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service