Skip to Content
Merck
All Photos(3)

Documents

197343

Sigma-Aldrich

Methyllithium solution

1.6 M in diethyl ether

Synonym(s):

Lithium methanide, MeLi

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3Li
CAS Number:
Molecular Weight:
21.98
Beilstein:
3587162
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

3 (vs air)

Quality Level

form

liquid

composition

halide, ~0.05 M

concentration

1.6 M in diethyl ether

density

0.732 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Li]C

InChI

1S/CH3.Li/h1H3;

InChI key

DVSDBMFJEQPWNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyllithium is an organolithium reagent mainly used for deprotonation and as a source of methyl anion. It is a strong base and a strong nucleophile.

Application

Methyllithium solution (1.6M in diethyl ether) has been used for the asymmetric allylic alkylation (AAA) of allylic electrophiles in the presence of a chiral copper catalyst. This protocol leads to the C-C bond formation of tertiary and quaternary stereogenic centers with high enantioselectivity. It can also be used for the synthesis of (−)-salsolidine by reacting with 6,7-dimethoxy-3,4-dihydroisoquinoline in the presence of (−)-sparteine as a chiral ligand.

Packaging

The 25 mL Sure/Seal bottle is recommended as a single-use bottle. Repeated punctures will likely result in decreased performance of product.

Legal Information

Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective addition of organolithium reagents to 3, 4-dihydroisoquinoline in the presence of (?)-sparteine as an external ligand. Application for the synthesis of isoquinoline alkaloids.
Chrzanowska M
Tetrahedron Asymmetry, 12(10), 1435-1440 (2001)
A Simplified Approach with 3D Visuals
Vasishta Bhatt
Essentials of Coordination Chemistry: A Simplified Approach with 3D Visuals, 183-183 (2015)
Mingyang Zhang et al.
Macromolecular rapid communications, 41(18), e2000323-e2000323 (2020-08-11)
Carbon monoxide (CO) has emerged as a potential therapeutic agent for the treatment of many diseases. However, the therapeutic outcome is highly dependent on the dosages and administration sites. Hence, there is mounting interest in the development of CO-releasing materials
Kiran Kumar Solingapuram Sai et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 129, 57-61 (2017-08-15)
We automated radiochemical synthesis of 1-[
Cu-catalyzed enantioselective allylic alkylation with organolithium reagents.
Hornillos V, et al.
Nature Protocols, 12(3), 493-493 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service