Merck
Bioelectronics
Brain illustrated as electronic chip surrounded by neural system to illustrate combination of state-of-the-art technology with bio-organisms.
Bioelectronics is a multidisciplinary field which integrates biology and electronics for diagnostic and therapeutic healthcare treatments. The regulatory activities of the nervous system can be monitored and controlled with bioelectronic technologies that affect specific molecular processes in neural signaling. Bioelectronic medicine manages a broad spectrum of diseases and disabilities, such as blindness, cardiovascular disease, diabetes, inflammatory and neurodegenerative diseases, and paralysis.  
Featued Categories
Representative chemical structure of polysiloxane, commonly known as silicone and silicone oil
Carbon Nanomaterials
A comprehensive offering of carbon nanomaterials from fullerene, to carbon nanotubes, graphene, carbon quantum dots and nanodiamonds to empower your research and development in energy storage, high performing electronics, smart composites, and novel nano therapeutics. 
Shop Products
Representative chemical structure of polysiloxane, commonly known as silicone and silicone oil
Silicone Materials
We offer a range of silicone materials with different molecular weights, chemical substitutes and terminal groups for use in soft lithography, surface modification, ink formulation, electronic packaging, thermal management, and biomedical applications.
Shop Products
Self-Assembly & Contact Printing
Self-Assembly & Contact Printing
We offer a comprehensive portfolio of self-assembly and nanoimprinting materials for effective target surface modification and precise transferal of micropatterns and nanopatterns to enable high-performance micro- and nanoelectronics.
Shop Products
Photonic & Optical Materials
Photonic & Optical Materials
Light up your research with our comprehensive portfolio of photonic and optical materials, including laser, infrared, near-infrared, chemiluminescent, photochromic, thermochromic, phthalocyanine, and porphyrin dyes, as well as photoluminescent, non-linear optical (NLO) and waveguide materials.
Shop Products
Bioelectronic devices function by stimulating, regulating, or even blocking specific electronic communication signals between the brain and bodily functions for personalized medical treatment. Common bioelectronic devices include cardiac pacemakers that modulate heart rate and robotic prostheses that emulate human movement. Biosensors, such as blood glucose monitors, detect enzymes, pathogens, or toxic substances. Wearable bioelectronic devices are capable of monitoring vital signs, detecting biomarkers, or harvesting epidermal energy. State-of-the-art bioelectronic implants can be powered without wires or batteries, and are minimally invasive, ingestible, and fully resorbable depending on the application.
Innovations in material design and device configurations have significantly improved the sensitivity and biocompatibility of bioelectronics. Soft, lightweight and ultra-thin materials, such as carbon nanotubes, graphene and other 2D nanomaterials, conductive polymers, gold nanoparticles, and quantum dots, are well suited for bioelectronic applications due to their excellent conductivity, flexibility, and miniature size.
For more information, read our Material Matters issue dedicated to bioelectronics.
Document Search
Looking for More Specific Information?
Visit our document search for data sheets, certificates and technical documentation.
Find Documents