Merck
HPLC Analysis of Proteins with Different pI Values on Proteomix® SCX-NP3
HPLC Analysis of Proteins with Different pI Values on Proteomix® SCX-NP3 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Proteomix® SCX-NP3 Strong Cation Exchange Column
L × I.D. 5 cm × 4.6 mm, 3 μm particle size, stainless steel
standard
Product No.
Description
Pricing
Albumin from chicken egg white
lyophilized powder, ≥98% (agarose gel electrophoresis)
Lysozyme from chicken egg white
powder or granules, ≥39,000 units/mg protein
Aprotinin from bovine lung
lyophilized powder, 3-8 TIU/mg solid
Ribonuclease A from bovine pancreas
Molecular Biology, ≥70 Kunitz units/mg protein, lyophilized
CONDITIONS
column
Proteomix SCX-NP3, 5 cm x 4.6 mm I.D., 3 μm particles (Z777151)
mobile phase
(A) 10 mM sodium phosphate, pH 6.0; (B) A + 1 M NaCl
gradient
0 to 100% B in 15 min
flow rate
0.5 mL/min
detector
UV, 280 nm
Description
Legal Information
Proteomix is a registered trademark of Sepax Technologies