GC Analysis of 5-Amino-1,3,3-Trimethylcyclohexanemethylamine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX™ G-TA
CONDITIONS
column
Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 μm (73033AST)
oven
160 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 30 psi
injection
1 μL, 80:1 split
sample gas
3 mg/mL racemate in methanol
peaks 1 & 2: first enantiomer pair
peaks 3 & 4: second enantiomer pair
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC