Merck
  • Controlling transgene expression in subcutaneous implants using a skin lotion containing the apple metabolite phloretin.
Controlling transgene expression in subcutaneous implants using a skin lotion containing the apple metabolite phloretin.
Proceedings of the National Academy of Sciences of the United States of America (2009-06-25)
Marc Gitzinger, Christian Kemmer, Marie Daoud El-Baba, Wilfried Weber, Martin Fussenegger
ABSTRACT
Adjustable control of therapeutic transgenes in engineered cell implants after transdermal and topical delivery of nontoxic trigger molecules would increase convenience, patient compliance, and elimination of hepatic first-pass effect in future therapies. Pseudomonas putida DOT-T1E has evolved the flavonoid-triggered TtgR operon, which controls expression of a multisubstrate-specific efflux pump (TtgABC) to resist plant-derived defense metabolites in its rhizosphere habitat. Taking advantage of the TtgR operon, we have engineered a hybrid P. putida-mammalian genetic unit responsive to phloretin. This flavonoid is contained in apples, and, as such, or as dietary supplement, regularly consumed by humans. The engineered mammalian phloretin-adjustable control element (PEACE) enabled adjustable and reversible transgene expression in different mammalian cell lines and primary cells. Due to the short half-life of phloretin in culture, PEACE could also be used to program expression of difficult-to-produce protein therapeutics during standard bioreactor operation. When formulated in skin lotions and applied to the skin of mice harboring transgenic cell implants, phloretin was able to fine-tune target genes and adjust heterologous protein levels in the bloodstream of treated mice. PEACE-controlled target gene expression could foster advances in biopharmaceutical manufacturing as well as gene- and cell-based therapies.
Sigma-Aldrich
Phloretin, ≥99%
Sigma-Aldrich
ω-Agatoxin TK, ~98% (HPLC), powder
Sigma-Aldrich
2-Naphthol, 99%
Sigma-Aldrich
(±)-Naringenin, ≥95%
Sigma-Aldrich
Phloridzin dihydrate, from apple wood, ≥99% (HPLC)