Merck
  • Prostate-specific membrane antigen cleavage of vitamin B9 stimulates oncogenic signaling through metabotropic glutamate receptors.
Prostate-specific membrane antigen cleavage of vitamin B9 stimulates oncogenic signaling through metabotropic glutamate receptors.
The Journal of experimental medicine (2017-11-17)
Charalambos Kaittanis, Chrysafis Andreou, Haley Hieronymus, Ninghui Mao, Catherine A Foss, Matthias Eiber, Gregor Weirich, Palak Panchal, Anuradha Gopalan, Juan Zurita, Samuel Achilefu, Gabriela Chiosis, Vladimir Ponomarev, Markus Schwaiger, Brett S Carver, Martin G Pomper, Jan Grimm
ABSTRACT
Prostate-specific membrane antigen (PSMA) or folate hydrolase 1 (FOLH1) is highly expressed on prostate cancer. Its expression correlates inversely with survival and increases with tumor grade. However, the biological role of PSMA has not been explored, and its role in prostate cancer remained elusive. Filling this gap, we demonstrate that in prostate cancer, PSMA initiates signaling upstream of PI3K through G protein-coupled receptors, specifically via the metabotropic glutamate receptor (mGluR). PSMA's carboxypeptidase activity releases glutamate from vitamin B9 and other glutamated substrates, which activate mGluR I. Activated mGluR I subsequently induces activation of phosphoinositide 3-kinase (PI3K) through phosphorylation of p110β independent of
Sigma-Aldrich
2-Isocyanatoethyl methacrylate, contains ≤0.1% BHT as inhibitor, 98%
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid
Sigma-Aldrich
L-(+)-2-Amino-3-phosphonopropionic acid
Sigma-Aldrich
Quisqualic acid, powder