Merck
  • Metabolomic Analysis Reveals Vitamin D-induced Decrease in Polyol Pathway and Subtle Modulation of Glycolysis in HEK293T Cells.
Metabolomic Analysis Reveals Vitamin D-induced Decrease in Polyol Pathway and Subtle Modulation of Glycolysis in HEK293T Cells.
Scientific reports (2017-08-27)
G C Santos, J D Zeidler, J A Pérez-Valencia, A C B Sant'Anna-Silva, A T Da Poian, T El-Bacha, F C L Almeida
ABSTRACT
We combined
Sigma-Aldrich
Ammonia Assay Kit, sufficient for 100 assays