Merck
  • Gastrointestinal Inhibition of Sodium-Hydrogen Exchanger 3 Reduces Phosphorus Absorption and Protects against Vascular Calcification in CKD.
Gastrointestinal Inhibition of Sodium-Hydrogen Exchanger 3 Reduces Phosphorus Absorption and Protects against Vascular Calcification in CKD.
Journal of the American Society of Nephrology : JASN (2014-11-19)
Eric D Labonté, Christopher W Carreras, Michael R Leadbetter, Kenji Kozuka, Jill Kohler, Samantha Koo-McCoy, Limin He, Edward Dy, Deborah Black, Ziyang Zhong, Ingrid Langsetmo, Andrew G Spencer, Noah Bell, Desiree Deshpande, Marc Navre, Jason G Lewis, Jeffrey W Jacobs, Dominique Charmot
ABSTRACT
In CKD, phosphate retention arising from diminished GFR is a key early step in a pathologic cascade leading to hyperthyroidism, metabolic bone disease, vascular calcification, and cardiovascular mortality. Tenapanor, a minimally systemically available inhibitor of the intestinal sodium-hydrogen exchanger 3, is being evaluated in clinical trials for its potential to (1) lower gastrointestinal sodium absorption, (2) improve fluid overload-related symptoms, such as hypertension and proteinuria, in patients with CKD, and (3) reduce interdialytic weight gain and intradialytic hypotension in ESRD. Here, we report the effects of tenapanor on dietary phosphorous absorption. Oral administration of tenapanor or other intestinal sodium-hydrogen exchanger 3 inhibitors increased fecal phosphorus, decreased urine phosphorus excretion, and reduced [(33)P]orthophosphate uptake in rats. In a rat model of CKD and vascular calcification, tenapanor reduced sodium and phosphorus absorption and significantly decreased ectopic calcification, serum creatinine and serum phosphorus levels, circulating phosphaturic hormone fibroblast growth factor-23 levels, and heart mass. These results indicate that tenapanor is an effective inhibitor of dietary phosphorus absorption and suggest a new approach to phosphate management in renal disease and associated mineral disorders.
Sigma-Aldrich
Anti-Na+/H+ Exchanger-3 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-Actin antibody produced in mouse, clone AC-40, ascites fluid