Merck
  • Differentiation Potential of Adipose-Derived Stem Cells When Cocultured with Smooth Muscle Cells, and the Role of Low-Intensity Laser Irradiation.
Differentiation Potential of Adipose-Derived Stem Cells When Cocultured with Smooth Muscle Cells, and the Role of Low-Intensity Laser Irradiation.
Photomedicine and laser surgery (2015-11-26)
Bernard Mvula, Heidi Abrahamse
ABSTRACT
The aim of the study was to investigate the differentiation potential of adipose-derived stem cells (ADSCs) when cocultured with smooth muscle cells (SMCs), and to determine the role of low-intensity laser irradiation (LILI). ADSCs isolated from adipose tissue are isolated with ease and in large amounts. SMCs constitute most parts of the intestinal, urinary, reproductive, and cardiovascular systems. LILI has been found to have positive effects on different cell types, including ADSCs. The study used ADSCs (Stempro Adipose Derived Stem Cells-R7788-115) and SMCs (SKU-T-1 American Type Culture Collection HTB-114) cell lines. These cell lines were cocultured in a 1:1 ratio with and without growth factors and then exposed to LILI using 636 nm at 5 J/cm Cell viability and proliferation increased significantly in the cocultured groups that were exposed to LILI alone, as well as in combination with growth factors. Further, there was a significant decrease in the expression of stem cell markers with a concomitant increase in SMC markers. These results suggest that ADSCs have the ability to differentiate into SMCs when cocultured with SMCs, whereas LILI potentially augments the differentiation potential and need. This further highlights the significant role that LILI has to offer ADSC therapy in regenerative medicine.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine and sodium bicarbonate, without HEPES, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, microbiologically tested, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline