Merck
  • Abundant DNA 6mA methylation during early embryogenesis of zebrafish and pig.
Abundant DNA 6mA methylation during early embryogenesis of zebrafish and pig.
Nature communications (2016-10-08)
Jianzhao Liu, Yuanxiang Zhu, Guan-Zheng Luo, Xinxia Wang, Yanan Yue, Xiaona Wang, Xin Zong, Kai Chen, Hang Yin, Ye Fu, Dali Han, Yizhen Wang, Dahua Chen, Chuan He
ABSTRACT
DNA N6-methyldeoxyadenosine (6mA) is a well-known prokaryotic DNA modification that has been shown to exist and play epigenetic roles in eukaryotic DNA. Here we report that 6mA accumulates up to ∼0.1-0.2% of total deoxyadenosine during early embryogenesis of vertebrates, but diminishes to the background level with the progression of the embryo development. During this process a large fraction of 6mAs locate in repetitive regions of the genome.
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC)
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000