Merck
  • Broadband photocurrent enhancement and light-trapping in thin film Si solar cells with periodic Al nanoparticle arrays on the front.
Broadband photocurrent enhancement and light-trapping in thin film Si solar cells with periodic Al nanoparticle arrays on the front.
Optics express (2015-06-16)
C Uhrenfeldt, T F Villesen, A Têtu, B Johansen, A Nylandsted Larsen
ABSTRACT
Plasmonic resonances in metal nanoparticles are considered candidates for improved thin film Si photovoltaics. In periodic arrays the influence of collective modes can enhance the resonant properties of such arrays. We have investigated the use of periodic arrays of Al nanoparticles placed on the front of a thin film Si test solar cell. It is demonstrated that the resonances from the Al nanoparticle array causes a broadband photocurrent enhancement ranging from the ultraviolet to the infrared with respect to a reference cell. From the experimental results as well as from numerical simulations it is shown that this broadband enhancement is due to single particle resonances that give rise to light-trapping in the infrared spectral range and to collective resonances that ensure an efficient in-coupling of light in the ultraviolet-blue spectral range.
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Silicon dioxide, single crystal substrate, optical grade, 99.99% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Silicon dioxide, granular, ≥99.9%
Sigma-Aldrich
Silicon dioxide, fused (granular), 4-20 mesh, 99.9% trace metals basis
Sigma-Aldrich
Silicon dioxide, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Supelco
Silica, 99.8%
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silica
Supelco
Glass spheres
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Silica, fumed, powder, 0.2-0.3 μm avg. part. size (aggregate)
Sigma-Aldrich
1-Naphthyl phosphate monosodium salt monohydrate, ≥98% (titration), powder
Sigma-Aldrich
Silica, fumed, powder
Sigma-Aldrich
Silica, mesostructured, SBA-15, 99% trace metals basis
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Sand, white quartz, ≥99.995% trace metals basis
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O