Merck
  • Sequential delivery of BMP-2 and BMP-7 for bone regeneration using a heparinized collagen membrane.
Sequential delivery of BMP-2 and BMP-7 for bone regeneration using a heparinized collagen membrane.
International journal of oral and maxillofacial surgery (2015-03-15)
J-Y Jo, S-I Jeong, Y-M Shin, S-S Kang, S-E Kim, C-M Jeong, J-B Huh
ABSTRACT
To investigate the effect of the sequential delivery of bone morphogenetic proteins BMP-2 and BMP-7 on bone regeneration in rat calvarial defects (40 Sprague-Dawley rats, 8mm defect size), all animals were treated with a hydroxyapatite (HA)/tricalcium phosphate (TCP) bone graft covered with a collagen membrane. The experimental groups were as follows: (1) control group: unmodified collagen (no treatment); (2) BMP-2 group: 5 μg of BMP-2; (3) hep-BMP-7 group: 5 μg BMP-7 chemically bound to heparinized collagen; and (4) BMP-2/hep-BMP-7 group: 2.5 μg BMP-7 bound to heparinized collagen and subsequently treated with 2.5 μg BMP-2. Defect healing was examined at 2 and 8 weeks after surgery. The BMP-2 group showed the largest new bone area at week 2 (29.3 ± 7.3%; P = 0.009); new bone areas in the hep-BMP-7 and BMP-2/hep-BMP-7 groups were similar (11.8 ± 3.4% and 12.9 ± 5.71%, respectively; P = 0.917). After 8 weeks, the BMP-2/hep-BMP-7 group showed the largest new bone area (43.3 ± 6.2%), followed by the BMP-2 and hep-BMP-7 groups (P = 0.013). Accordingly, in comparison with single deliveries of BMP-2 and BMP-7, sequential delivery of BMP-2 and BMP-7 using a heparinized collagen membrane significantly induced new bone formation with a smaller quantity of BMP-2 in rat calvarial defects.
Sigma-Aldrich
4-Ethylmorpholine, 97%
Sigma-Aldrich
4-Ethylmorpholine, BioXtra, suitable for protein sequencing, ≥99.5% (GC)
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%