Merck
  • Phase I Study of Vorinostat as a Radiation Sensitizer with 131I-Metaiodobenzylguanidine (131I-MIBG) for Patients with Relapsed or Refractory Neuroblastoma.
Phase I Study of Vorinostat as a Radiation Sensitizer with 131I-Metaiodobenzylguanidine (131I-MIBG) for Patients with Relapsed or Refractory Neuroblastoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2015-02-20)
Steven G DuBois, Susan Groshen, Julie R Park, Daphne A Haas-Kogan, Xiaodong Yang, Ethan Geier, Eugene Chen, Kathy Giacomini, Brian Weiss, Susan L Cohn, M Meaghan Granger, Gregory A Yanik, Randall Hawkins, Jesse Courtier, Hollie Jackson, Fariba Goodarzian, Hiroyuki Shimada, Scarlett Czarnecki, Denice Tsao-Wei, Judith G Villablanca, Araz Marachelian, Katherine K Matthay
ABSTRACT
(131)I-metaiodobenzylguanidine (MIBG) is a radiopharmaceutical with activity in neuroblastoma. Vorinostat is a histone deacetylase inhibitor that has radiosensitizing properties. The goal of this phase I study was to determine the MTDs of vorinostat and MIBG in combination. Patients ≤ 30 years with relapsed/refractory MIBG-avid neuroblastoma were eligible. Patients received oral vorinostat (dose levels 180 and 230 mg/m(2)) daily days 1 to 14. MIBG (dose levels 8, 12, 15, and 18 mCi/kg) was given on day 3 and peripheral blood stem cells on day 17. Alternating dose escalation of vorinostat and MIBG was performed using a 3+3 design. Twenty-seven patients enrolled to six dose levels, with 23 evaluable for dose escalation. No dose-limiting toxicities (DLT) were seen in the first three dose levels. At dose level 4 (15 mCi/kg MIBG/230 mg/m(2) vorinostat), 1 of 6 patients had DLT with grade 4 hypokalemia. At dose level 5 (18 mCi/kg MIBG/230 mg/m(2) vorinostat), 2 patients had dose-limiting bleeding (one grade 3 and one grade 5). At dose level 5a (18 mCi/kg MIBG/180 mg/m(2) vorinostat), 0 of 6 patients had DLT. The most common toxicities were neutropenia and thrombocytopenia. The response rate was 12% across all dose levels and 17% at dose level 5a. Histone acetylation increased from baseline in peripheral blood mononuclear cells collected on days 3 and 12 to 14. Vorinostat at 180 mg/m(2)/dose is tolerable with 18 mCi/kg MIBG. A phase II trial comparing this regimen to single-agent MIBG is ongoing.
Sigma-Aldrich
m-Iodobenzylguanidine hemisulfate salt, ≥98% (HPLC and TLC)
Pricing and availability is not currently available.