Merck
  • Identification of 14-3-3 proteins in human platelets: effects of synthetic peptides on protein kinase C activation.
Identification of 14-3-3 proteins in human platelets: effects of synthetic peptides on protein kinase C activation.
The Biochemical journal (1996-04-01)
C P Wheeler-Jones, M P Learmonth, H Martin, A Aitken
ABSTRACT
The 14-3-3 proteins inhibit protein kinase C (PKC) activity in vitro and contain conserved sequences that resemble the pseudosubstrate domain of PKC and the C-terminus of the annexins. In the present study we have identified the isoforms of 14-3-3 in human platelets and used synthetic peptides derived from the regions with similarity to PKC and annexins to examine the potential role of 14-3-3 in regulating platelet activity. Immunoblotting studies with isoform-specific antisera raised against the acetylated peptides corresponding to the N-termini of 14-3-3 showed that these cells express high levels of the beta, gamma and zeta 14-3-3 isoforms. In addition, low levels of the epsilon and eta 14-3-3 isoforms were detected. In washed, saponin-permeabilized platelets incubated with [gamma-32P]ATP, thrombin- and phorbol 12-myristate 13-acetate (PMA)-induced phosphorylation of several proteins (66, 45, and 20kDa) was inhibited by preincubation with AS peptide (KNVVGARRSSWRVISSIEQK) based on the pseudosubstrate-like region of the 14-3-3 family. A control peptide of similar size had no effect on PKC-mediated phosphorylation. PMA caused a rapid translocation of PKC activity from the cytosol to the particulate fraction of saponin-permeabilized platelets that was unaffected by either the AS peptide or a peptide derived from the annexin-like 14-3-3 domain (MKGDYYRYLAEVATGDD). These results suggest that isoforms of the 14-3-3 family may play an important physiological role as inhibitors of PKC activity in human platelets but are unlikely to be involved in controlling association of PKC with the membrane.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Sigma-Aldrich
Thrombin human, BioUltra, recombinant, expressed in HEK 293 cells, aqueous solution, ≥95% (SDS-PAGE)
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
USP
Epoprostenol sodium, United States Pharmacopeia (USP) Reference Standard