Merck
  • Direct separation of the diastereomers of phosphatidylcholine hydroperoxide bearing 13-hydroperoxy-9Z,11E-octadecadienoic acid using chiral stationary phase high-performance liquid chromatography.
Direct separation of the diastereomers of phosphatidylcholine hydroperoxide bearing 13-hydroperoxy-9Z,11E-octadecadienoic acid using chiral stationary phase high-performance liquid chromatography.
Journal of chromatography. A (2015-02-18)
Junya Ito, Kiyotaka Nakagawa, Shunji Kato, Takafumi Hirokawa, Shigefumi Kuwahara, Toshiharu Nagai, Teruo Miyazawa
ABSTRACT
Increasing evidence suggests that phospholipid peroxidation plays important roles in the pathogenesis of various diseases, such as atherosclerosis. With regard to the biochemical processes that initiate phospholipid peroxidation in vivo, enzymatic conversion of phosphatidylcholine to phosphatidylcholine hydroperoxide (PCOOH) by lipoxygenase (LOX) may play a crucial role. This will become clear if we can analyze PCOOH bearing hydroperoxy fatty acids with S-stereoconfiguration. In this study, we therefore attempted such an analysis. Initially, we used LOX, linoleic acid and Lyso phosphatidylcholine, and synthesized PCOOH bearing 13S-hydroperoxy-9Z,11E-octadecadienoic acid (13(S)-9Z,11E-HPODE). PCOOH bearing racemic 13-9Z,11E-HPODE was also prepared. We used liquid chromatography equipped with CHIRALPAK OP (+) (poly (o-pyridyl diphenylmethacrylate) coated on silica), a UV detector and a quadrupole-time-of-flight mass spectrometer, and achieved diastereomer separation of PCOOH stereoisomers with excellent resolution and peak shape. This is the first study reporting the diastereomer separation of PCOOH. The present method will be beneficial in developing a better understanding of the biochemical processes that initiate oxidative stress (PCOOH formation) in vivo, which may lead to further elucidation of the involvement of PCOOH in the development of diseases. In addition to clinical applications, the present method may also be effective in the evaluation of enzymatic oxidative food deterioration.
Sigma-Aldrich
Pyridinium p-toluenesulfonate, puriss., ≥99.0% (T)
Sigma-Aldrich
2-Methoxypropene, 97%
Valaciclovir impurity G, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Pyridinium p-toluenesulfonate, 98%
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
4-(Dimethylamino)pyridine, purum, ≥98.0% (NT)
Sigma-Aldrich
4-(Dimethylamino)pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%