Merck
  • LOX-1 - dependent mitochondrial DNA damage and NLRP3 activation during systemic inflammation in mice.
LOX-1 - dependent mitochondrial DNA damage and NLRP3 activation during systemic inflammation in mice.
Biochemical and biophysical research communications (2014-08-19)
Zufeng Ding, Shijie Liu, Xianwei Wang, Sue Theus, Yubo Fan, Xiaoyan Deng, Jawahar L Mehta
ABSTRACT
Lectin-like oxidized low-density lipoprotein scavenger receptor-1 (LOX-1) is known to be involved in many pathophysiological events, such as inflammation. To clarify the role of LOX-1 in mtDNA damage and NLRP3 inflammasome activation, we studied wild-type (WT) and LOX-1 knockout (KO) mice given thioglycollate, an inflammatory stimulus. We observed intense inflammatory response (CD45 and CD68 expression) and mtDNA damage in spleen and kidneys of WT mice given thioglycollate. The abrogation of LOX-1 (use of LOX-1 knockout mice) reduced the inflammatory response as well as mtDNA damage (P<0.05 vs. WT mice). We also observed that mice with LOX-1 deletion had markedly reduced expression of caspase-1 (P10 and P20 subunits) as well as cleaved IL-1β and IL-18. These mice also had much less mtDNA damage and only limited NLRP3 inflammasome expression. These in vivo observations indicate that LOX-1 plays a key role in mtDNA damage which then leads to NLRP3 inflammasome activation during inflammation.
Sigma-Aldrich
Mouse Il18 / Interleukin-18 ELISA Kit, for serum, plasma and cell culture supernatants
Sigma-Aldrich
Human IL-18 BPa  ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IL-18 R β ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Human IL18 / Interleukin-18 ELISA Kit, for serum, plasma, cell culture supernatants and urine