Merck
  • Activation of HIV transcription with short-course vorinostat in HIV-infected patients on suppressive antiretroviral therapy.
Activation of HIV transcription with short-course vorinostat in HIV-infected patients on suppressive antiretroviral therapy.
PLoS pathogens (2014-11-14)
Julian H Elliott, Fiona Wightman, Ajantha Solomon, Khader Ghneim, Jeffrey Ahlers, Mark J Cameron, Miranda Z Smith, Tim Spelman, James McMahon, Pushparaj Velayudham, Gregor Brown, Janine Roney, Jo Watson, Miles H Prince, Jennifer F Hoy, Nicolas Chomont, Rémi Fromentin, Francesco A Procopio, Joumana Zeidan, Sarah Palmer, Lina Odevall, Ricky W Johnstone, Ben P Martin, Elizabeth Sinclair, Steven G Deeks, Daria J Hazuda, Paul U Cameron, Rafick-Pierre Sékaly, Sharon R Lewin
ABSTRACT
Human immunodeficiency virus (HIV) persistence in latently infected resting memory CD4+ T-cells is the major barrier to HIV cure. Cellular histone deacetylases (HDACs) are important in maintaining HIV latency and histone deacetylase inhibitors (HDACi) may reverse latency by activating HIV transcription from latently infected CD4+ T-cells. We performed a single arm, open label, proof-of-concept study in which vorinostat, a pan-HDACi, was administered 400 mg orally once daily for 14 days to 20 HIV-infected individuals on suppressive antiretroviral therapy (ART). The primary endpoint was change in cell associated unspliced (CA-US) HIV RNA in total CD4+ T-cells from blood at day 14. The study is registered at ClinicalTrials.gov (NCT01365065). Vorinostat was safe and well tolerated and there were no dose modifications or study drug discontinuations. CA-US HIV RNA in blood increased significantly in 18/20 patients (90%) with a median fold change from baseline to peak value of 7.4 (IQR 3.4, 9.1). CA-US RNA was significantly elevated 8 hours post drug and remained elevated 70 days after last dose. Significant early changes in expression of genes associated with chromatin remodeling and activation of HIV transcription correlated with the magnitude of increased CA-US HIV RNA. There were no statistically significant changes in plasma HIV RNA, concentration of HIV DNA, integrated DNA, inducible virus in CD4+ T-cells or markers of T-cell activation. Vorinostat induced a significant and sustained increase in HIV transcription from latency in the majority of HIV-infected patients. However, additional interventions will be needed to efficiently induce virus production and ultimately eliminate latently infected cells. ClinicalTrials.gov NCT01365065.
Product Description
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder