Merck
  • Replication rates of Mycobacterium tuberculosis in human macrophages do not correlate with mycobacterial antibiotic susceptibility.
Replication rates of Mycobacterium tuberculosis in human macrophages do not correlate with mycobacterial antibiotic susceptibility.
PloS one (2014-11-12)
Johanna Raffetseder, Elsje Pienaar, Robert Blomgran, Daniel Eklund, Veronika Patcha Brodin, Henrik Andersson, Amanda Welin, Maria Lerm
ABSTRACT
The standard treatment of tuberculosis (TB) takes six to nine months to complete and this lengthy therapy contributes to the emergence of drug-resistant TB. TB is caused by Mycobacterium tuberculosis (Mtb) and the ability of this bacterium to switch to a dormant phenotype has been suggested to be responsible for the slow clearance during treatment. A recent study showed that the replication rate of a non-virulent mycobacterium, Mycobacterium smegmatis, did not correlate with antibiotic susceptibility. However, the question whether this observation also holds true for Mtb remains unanswered. Here, in order to mimic physiological conditions of TB infection, we established a protocol based on long-term infection of primary human macrophages, featuring Mtb replicating at different rates inside the cells. During conditions that restricted Mtb replication, the bacterial phenotype was associated with reduced acid-fastness. However, these phenotypically altered bacteria were as sensitive to isoniazid, pyrazinamide and ethambutol as intracellularly replicating Mtb. In support of the recent findings with M. smegmatis, we conclude that replication rates of Mtb do not correlate with antibiotic tolerance.
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
2-Methyl-2-propanethiol, 99%
Sigma-Aldrich
Decanal, ≥98% (GC), liquid
SAFC
L-Glutamine
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine
Supelco
Decanal, analytical standard
Sigma-Aldrich
Decanal, FCC, FG
Sigma-Aldrich
Decanal, natural, ≥97%, FG