Merck
  • Lipid peroxidation, DNA damage and coenzyme Q10 in lung cancer patients--markers for risk assessment?
Lipid peroxidation, DNA damage and coenzyme Q10 in lung cancer patients--markers for risk assessment?
Asian Pacific journal of cancer prevention : APJCP (2011-12-01)
Ufuk Cobanoglu, Halit Demir, Aysegul Cebi, Fuat Sayir, Hamit Hakan Alp, Zafer Akan, Tugba Gur, Ebubekir Bakan
ABSTRACT
Early diagnosis and prevention is very important for lung cancer patients. Previous studies have emphasized that the level of coenzyme Q10 (CoQ10), present primarily in mitochondria, decreases with age and is low in patients with chronic diseases. Our goal was to find out if there is any relationship between lung cancer and CoQ10 and lipid peroxidation levels. Blood samples from lung cancer patients were collected. Total and oxide CoQ10 levels, 8-OHdG (product of DNA damage), and malondialdehyde (MDA) levels (lipid peroxidation) were analyzed with high performance liquid chromatography (HPLC). The MDA level (P<0.001) and DNA damage rate (8-OHdG) (P<0.001) was higher in cancer patients than in the control group; in contrast, theCoQ10 enzyme level was significantly lower (P<0.001). The results suggest that the aforementioned parameters can be useful for lung cancer risk assessment.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Coenzyme Q10, ≥98% (HPLC)
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, ≥99.9%