Merck
High resolution 31P NMR of extracted phospholipids.
Journal of lipid research (1988-05-01)
P Meneses, T Glonek
ABSTRACT
The common phospholipids from biological sources were quantitated using phosphorus-31 nuclear magnetic resonance (NMR) spectroscopy in conjunction with an analytical reagent composed of two parts: 1) 2 ml of reagent chloroform in which was dissolved 0.01-100 mg of crude tissue lipid extracted from tissue sources using chloroform-methanol 2:1, the extract having been washed with 0.2 vol. of 0.1 M KCl; 2) 1 ml of an aqueous methanol reagent composed of one part 0.2 M (ethylenedinitrilo)-tetraacetic acid in D2O titrated to pH 6 with CsOH and four parts of reagent methanol. In a magnetic field of 11.75 Tesla, the extracted phospholipids yield narrow signals (1.8-3.2 Hz at half-height), corresponding to each generic species, e.g., phosphatidylcholines, phosphatidylethanolamines, etc., permitting resolution among the various phospholipid families and their lyso and plasmalogen derivatives. The reagent permits assays of high precision and accuracy using a modest amount of NMR spectrometer time (ca. 15 min/assay). The procedures described, which are compared to high-performance liquid chromatography, are convenient for the routine analysis of phospholipids from biological sources.
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
Phosphoric acid, puriss., ≥99%
Sigma-Aldrich
Phosphoric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O