Merck
  • RNA-binding proteins regulate the expression of the immune activating ligand MICB.
RNA-binding proteins regulate the expression of the immune activating ligand MICB.
Nature communications (2014-06-14)
Daphna Nachmani, Tony Gutschner, Adi Reches, Sven Diederichs, Ofer Mandelboim
ABSTRACT
The recognition of stress-induced ligands by the activating receptor NKG2D expressed on cytotoxic lymphocytes is crucial for the prevention and containment of various diseases and is also one of the best-studied examples of how danger is sensed by the immune system. Still, however, the mechanisms leading to the expression of the NKG2D ligands are far from being completely understood. Here, we use an unbiased and systematic RNA pull-down approach combined with mass spectrometry to identify six RNA-binding proteins (RBPs) that bind and regulate the expression of MICB, one of the major stress-induced ligands of NKG2D. We further demonstrate that at least two of the identified RBPs function during genotoxic stress. Our data provide insights into stress recognition and hopefully open new therapeutic venues.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
RIPAb+ HuR - RIP Validated Antibody and Primer Set, from rabbit
Sigma-Aldrich
Anti-CUGBP1, (N-terminal) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
Anti-HSP90 Antibody, clone D7a, Upstate®, from mouse
Sigma-Aldrich
Anti-c-Jun Antibody, serum, Upstate®
Sigma-Aldrich
RIPAb+ IGF2BP2 Antibody, clone 5E10-3E1, from mouse