Merck
  • Asymmetric palladium-catalysed intramolecular Wacker-type cyclisations of unsaturated alcohols and amino alcohols.
Asymmetric palladium-catalysed intramolecular Wacker-type cyclisations of unsaturated alcohols and amino alcohols.
Molecules (Basel, Switzerland) (2013-05-28)
Jana Doháňošová, Tibor Gracza
ABSTRACT
The palladium (II)-catalysed reactions of alkenols and aminoalkenols such as oxycarbonylations or bicyclisations are powerful methods for the construction of oxygen and nitrogen-containing heterocyclic compounds. This review highlights recent progress in the development of the asymmetric palladium(II)-catalysed Wacker-type cyclisations of unsaturated polyols and aminoalcohols. The scope, limitations, and applications of these reactions are presented.
Sigma-Aldrich
Palladium, powder or granules, 99.99% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Palladium, powder, <75 μm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, 99.995% trace metals basis
Sigma-Aldrich
Palladium, sponge, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 1.0 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.025 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, wire, diam. 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, foil, thickness 0.5 mm, 99.9% trace metals basis
Sigma-Aldrich
Palladium, powder, <1 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium, evaporation slug, diam. × L 0.6 cm × 0.6 cm, 99.95% trace metals basis
Sigma-Aldrich
Palladium, nanopowder, <25 nm particle size (TEM), ≥99.5%
Palladium, foil, 6mm disks, thickness 0.03mm, as rolled, 99.95%
Palladium, foil, 0.5m coil, thickness 0.1mm, coil width 2mm, as rolled, 99.95%
Palladium, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Palladium, foil, 4mm disks, thickness 0.50mm, as rolled, 99.95%
Palladium, foil, 10mm disks, thickness 0.125mm, as rolled, 99.95%
Palladium, foil, 4mm disks, thickness 0.025mm, as rolled, 99.95%
Palladium, foil, 15mm disks, thickness 0.03mm, as rolled, 99.95%
Palladium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Palladium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.95%
Palladium, foil, 15mm disks, thickness 0.047mm, as rolled, 99.95%
Palladium, foil, 10mm disks, thickness 0.25mm, as rolled, 99.99+%
Palladium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%
Palladium, foil, 25x25mm, thickness 0.50mm, as rolled, 99.95%
Palladium, foil, 4mm disks, thickness 0.02mm, as rolled, 99.95%
Palladium, foil, 15mm disks, thickness 0.075mm, as rolled, 99.95%
Palladium, foil, 10mm disks, thickness 0.0125mm, 99.95%
Palladium, foil, 10x10mm, thickness 0.50mm, as rolled, 99.95%