Merck
Phloroglucinol compounds of natural origin: synthetic aspects.
Natural product reports (2010-02-25)
Inder Pal Singh, Jasmeen Sidana, Sandip B Bharate, William J Foley
ABSTRACT
This review covers the synthetic aspects of various naturally occurring phloroglucinols, describing syntheses of 42 compounds and citing 137 references.
Sigma-Aldrich
Phloroglucinol, ≥99.0% (HPLC)
Sigma-Aldrich
Phloroglucinol, Used to detect the presence of wood fiber.
Sigma-Aldrich
Phloroglucinol, suitable for plant cell culture, BioReagent
Phloroglucinol (anhydrous), European Pharmacopoeia (EP) Reference Standard