- Cell biology of molybdenum.
ABSTRACT
The transition element molybdenum (Mo) is an essential micronutrient that is needed as catalytically active metal during enzyme catalysis. In humans four enzymes depend on Mo: sulfite oxidase, xanthine oxidoreductase, aldehyde oxidase, and mitochondrial amidoxime reductase. In addition to these enzymes, plants harbor a fifth Mo-enzyme namely nitrate reductase. To gain biological activity and fulfill its function in enzymes, Mo has to be complexed by a pterin compound thus forming the molybdenum cofactor. This article will review the way that Mo takes from uptake into the cell, via formation of the molybdenum cofactor and its storage, up to the final insertion of the molybdenum cofactor into apometalloenzymes.
Product Description
Molybdenum, foil, 6mm disks, thickness 0.075mm, annealed, 99.9%
Molybdenum, foil, 50mm disks, thickness 0.25mm, annealed, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.0125mm, 99.9%
Molybdenum, foil, 50mm disks, thickness 0.125mm, annealed, 99.9%
Molybdenum, foil, 50mm disks, thickness 0.30mm, annealed, 99.9%
Molybdenum, foil, 6mm disks, thickness 0.015mm, annealed, 99.9%
Molybdenum, rod, 100mm, diameter 12mm, centerless ground, 99.9%
Molybdenum, microfoil, disks, 25mm, thinness 1.0μm, specific density 1022μg/cm2, permanent mylar 3.5μm support, 99.9%
Molybdenum, foil, not light tested, 50x50mm, thickness 0.007mm, 99.9%
Molybdenum, pellets, 50g, max. size 10mm, 99.9%
Molybdenum, insulated wire, 10m, conductor diameter 0.125mm, insulation thickness 0.012mm, polyimide insulation, 99.95%
Molybdenum, wire reel, 10m, diameter 0.013mm, hard, 99.8+%
Molybdenum, microfoil, disks, 10mm, thinness 0.1μm, specific density 92.4μg/cm2, permanent mylar 3.5μm support, 99.9%
Molybdenum, wire reel, 100m, diameter 0.03mm, hard, 99.95%
Molybdenum, foil, 100x100mm, thickness 0.15mm, annealed, 99.9%
Molybdenum, foil, not light tested, 25x25mm, thickness 0.006mm, 99.9%
Molybdenum, foil, not light tested, 50x50mm, thickness 0.002mm, 99.9%
Molybdenum, wire reel, 100m, diameter 1.25mm, annealed, 99.95%
Molybdenum, wire reel, 50m, diameter 0.25mm, annealed, 99.95%
Molybdenum, wire reel, 1m, diameter 1.0mm, annealed, 99.95%
Molybdenum, foil, 100x100mm, thickness 0.20mm, annealed, 99.9%
Molybdenum, wire reel, 1m, diameter 0.013mm, hard, 99.8+%
Molybdenum, wire reel, 200m, diameter 0.30mm, annealed, 99.95%
Molybdenum, foil, 100x100mm, thickness 2.0mm, annealed, 99.9%
Molybdenum, wire reel, 50m, diameter 0.2mm, as drawn, 99.95%