Merck
  • Involvement of active site in self-association of proteins: a case of alpha-chymotrypsin.
Involvement of active site in self-association of proteins: a case of alpha-chymotrypsin.
Indian journal of biochemistry & biophysics (1992-04-01)
T Ramakrishna, M W Pandit
ABSTRACT
The self-association of proteolytic enzymes can be looked upon as an interesting possibility of the manifestation of enzyme-substrate complex. Hence the involvement of active site in such processes is a centre of investigation for many years. In the case of alpha-chymotrypsin, considerable controversy exists with regard to the involvement of active site of the enzyme in its self-association. A historical perspective of the problem and an overview of the available evidence, for and against, is presented and critically analysed. Despite contradicting observations, accumulated evidence indicates that His-57 and Ser-195 at the active site are involved, at least partially, in the self-association; a few other groups such as Tyr-146 and Met-192 are also involved in such processes.
Product Description
Chymotrypsin, BRP, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
α-Chymotrypsin from human pancreas, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, ≥40 units/mg protein, vial of 5 mg
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type II, lyophilized powder, ≥40 units/mg protein
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, suitable for protein sequencing, salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, Type I-S, essentially salt-free, lyophilized powder
Sigma-Aldrich
α-Chymotrypsin from bovine pancreas, (TLCK treated to inactivate residual tryspin activity), Type VII, essentially salt-free, lyophilized powder, ≥40 units/mg protein