Merck
Aspirin in patients undergoing noncardiac surgery.
The New England journal of medicine (2014-04-01)
P J Devereaux, Marko Mrkobrada, Daniel I Sessler, Kate Leslie, Pablo Alonso-Coello, Andrea Kurz, Juan Carlos Villar, Alben Sigamani, Bruce M Biccard, Christian S Meyhoff, Joel L Parlow, Gordon Guyatt, Andrea Robinson, Amit X Garg, Reitze N Rodseth, Fernando Botto, Giovanna Lurati Buse, Denis Xavier, Matthew T V Chan, Maria Tiboni, Deborah Cook, Priya A Kumar, Patrice Forget, German Malaga, Edith Fleischmann, Mohammed Amir, John Eikelboom, Richard Mizera, David Torres, C Y Wang, Tomas VanHelder, Pilar Paniagua, Otavio Berwanger, Sadeesh Srinathan, Michelle Graham, Laura Pasin, Yannick Le Manach, Peggy Gao, Janice Pogue, Richard Whitlock, André Lamy, Clive Kearon, Colin Baigent, Clara Chow, Shirley Pettit, Susan Chrolavicius, Salim Yusuf
ABSTRACT
There is substantial variability in the perioperative administration of aspirin in patients undergoing noncardiac surgery, both among patients who are already on an aspirin regimen and among those who are not. Using a 2-by-2 factorial trial design, we randomly assigned 10,010 patients who were preparing to undergo noncardiac surgery and were at risk for vascular complications to receive aspirin or placebo and clonidine or placebo. The results of the aspirin trial are reported here. The patients were stratified according to whether they had not been taking aspirin before the study (initiation stratum, with 5628 patients) or they were already on an aspirin regimen (continuation stratum, with 4382 patients). Patients started taking aspirin (at a dose of 200 mg) or placebo just before surgery and continued it daily (at a dose of 100 mg) for 30 days in the initiation stratum and for 7 days in the continuation stratum, after which patients resumed their regular aspirin regimen. The primary outcome was a composite of death or nonfatal myocardial infarction at 30 days. The primary outcome occurred in 351 of 4998 patients (7.0%) in the aspirin group and in 355 of 5012 patients (7.1%) in the placebo group (hazard ratio in the aspirin group, 0.99; 95% confidence interval [CI], 0.86 to 1.15; P=0.92). Major bleeding was more common in the aspirin group than in the placebo group (230 patients [4.6%] vs. 188 patients [3.8%]; hazard ratio, 1.23; 95% CI, 1.01, to 1.49; P=0.04). The primary and secondary outcome results were similar in the two aspirin strata. Administration of aspirin before surgery and throughout the early postsurgical period had no significant effect on the rate of a composite of death or nonfatal myocardial infarction but increased the risk of major bleeding. (Funded by the Canadian Institutes of Health Research and others; POISE-2 ClinicalTrials.gov number, NCT01082874.).
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material