Merck
  • New hydrazines with sulphonamidic structure: synthesis,characterization and biological activity.
New hydrazines with sulphonamidic structure: synthesis,characterization and biological activity.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi (2014-02-11)
Oana Maria Parasca, Florentina Lupascu, Cornelia Vasile, M Mares, V Nastasa, Lenuta Profire
ABSTRACT
Infections caused by bacterial species are common in immunocompromised patients and carry significant treatment costs and mortality. The emerging resistance of microorganisms to some synthetic antimicrobial agents makes it necessary to continue the research for new antimicrobial drugs. To design new sulphonamide compounds with potential antibacterial and antioxidant activity. New N-hydrazino acetyl-sulphonamides were prepared by condensation of some sulphonamides with chloroacetyl chloride and amination of intermediate compounds with hydrazine hydrate. The synthesized compounds were screened for their antibacterial activity against Gram positive (Klebsiellapneumoniae. Proteus vulgaris, Citrobacter freundii, Enterobacter cloacae, Staphylococcus aureus, Staphylococcus epidermidis, Enterococcus faecalis) and Gram negative bacterial strains (Pseudomonas aeruginosa, Escherichia coli). Some of them were found to have good antibacterial activity. The antioxidant activity of these compounds was also tested using different methods: total antioxidant capacity, reducing power, and DPPH radical scavenging activity. Chemical modulations performed on sulphonamide structure have a good influence on the biological activity of the synthesized compounds, especially on their antioxidant effects.
Sigma-Aldrich
Chloroacetyl chloride, purum, ≥99.0% (GC)