Merck
  • Efficacy of anti-inflammatory or antibiotic treatment in patients with non-complicated acute bronchitis and discoloured sputum: randomised placebo controlled trial.
Efficacy of anti-inflammatory or antibiotic treatment in patients with non-complicated acute bronchitis and discoloured sputum: randomised placebo controlled trial.
BMJ (Clinical research ed.) (2013-10-08)
Carl Llor, Ana Moragas, Carolina Bayona, Rosa Morros, Helena Pera, Oleguer Plana-Ripoll, Josep M Cots, Marc Miravitlles
ABSTRACT
To evaluate the efficacy of oral anti-inflammatory or antibiotic treatment compared with placebo in the resolution of cough in patients with uncomplicated acute bronchitis and discoloured sputum. Multicentre, parallel, single blinded placebo controlled, randomised clinical trial. Nine primary care centres in Spain. Adults aged 18 to 70 presenting symptoms associated with respiratory tract infection of less than one week's duration, with cough as the predominant symptom, the presence of discoloured sputum, and at least one other symptom of lower respiratory tract infection (dyspnoea, wheezing, chest discomfort, or chest pain). Patients were randomised to receive either ibuprofen 600 mg three times daily, amoxicillin-clavulanic acid 500 mg/125 mg three times daily, or placebo three times daily for 10 days. The duration of symptoms was measured with a diary card. Number of days with frequent cough after the randomisation visit. 416 participants were randomised (136 to ibuprofen, 137 to antibiotic, and 143 to placebo) and 390 returned their symptom diaries fully completed. The median number of days with frequent cough was slightly lower among patients assigned to ibuprofen (9 days, 95% confidence interval 8 to 10 days) compared with those receiving amoxicillin-clavulanic acid (11 days, 10 to 12 days) or placebo (11 days, 8 to 14 days), albeit without statistically significant differences. Neither amoxicillin-clavulanic acid nor ibuprofen increased the probability of cough resolution (hazard ratio 1.03, 95% confidence interval 0.78 to 1.35 and 1.23, 0.93 to 1.61, respectively) compared with placebo. Adverse events were observed in 27 patients, and were more common in the antibiotic arm (12%) than ibuprofen or placebo arms (5% and 3%, respectively; P<0.01). No significant differences were observed in the number of days with cough between patients with uncomplicated acute bronchitis and discoloured sputum treated with ibuprofen, amoxicillin-clavulanic acid, or placebo. Current Controlled Trials ISRCTN07852892.
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Ibuprofen
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard