Merck
  • Scandium oxide coated polycrystalline tungsten studied using emission microscopy and photoelectron spectroscopy.
Scandium oxide coated polycrystalline tungsten studied using emission microscopy and photoelectron spectroscopy.
Ultramicroscopy (2011-11-15)
Congshang Wan, Joel M Vaughn, Jerzy T Sadowski, Martin E Kordesch
ABSTRACT
Thermionic electron emission from 200 to 500 nm thick coatings of scandium oxide on tungsten foil have been examined in thermionic emission microscopy, spectroscopic photoelectron microcopy, synchrotron radiation and ultraviolet photoelectron spectroscopy (UPS). A clear dependence of the scandium oxide-W electron yield on the grain orientation of the polycrystalline tungsten is observed in thermionic emission and photoelectron emission.
Product Description
Tungsten, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.5mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.04mm, coil width 0.7mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.05mm, coil width .51mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.125mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.2mm, as rolled, 99.95%
Tungsten, foil, 200x200mm, thickness 0.3mm, as rolled, 99.95%
Tungsten, foil, 200x200mm, thickness 0.15mm, as rolled, 99.95%
Tungsten, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95%
Tungsten, foil, 0.1m coil, thickness 0.076mm, coil width 2mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.125mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95%
Tungsten, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.025mm, coil width 1mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.05mm, coil width 1.5mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.2mm, as rolled, 99.95%
Tungsten, foil, 10mm disks, thickness 1.0mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.02mm, coil width .1mm, as rolled, 99.95%
Tungsten, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Tungsten, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.5mm, as rolled, 99.95%
Tungsten, foil, 25mm disks, thickness 1.0mm, as rolled, 99.95%
Tungsten, foil, 0.5m coil, thickness 0.02mm, coil width .1mm, as rolled, 99.95%
Tungsten, foil, 50x50mm, thickness 0.3mm, as rolled, 99.95%
Tungsten, foil, 1m coil, thickness 0.6mm, coil width 1mm, as rolled, 99.95%
Tungsten, foil, 2m coil, thickness 0.05mm, coil width .51mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.035mm, coil width .5mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.05mm, coil width 1.5mm, as rolled, 99.95%
Tungsten, foil, 2m coil, thickness 0.04mm, coil width 0.7mm, as rolled, 99.95%
Tungsten, foil, 0.2m coil, thickness 0.10mm, coil width .9mm, as rolled, 99.95%