Merck
Nanostructured thin solid oxide fuel cells with high power density.
Dalton transactions (Cambridge, England : 2003) (2008-12-17)
Alex Ignatiev, Xin Chen, Naijuan Wu, Zigui Lu, Laverne Smith
ABSTRACT
Nanostructured thin film solid oxide fuel cells (SOFC) have been developed for reduced temperature operation, with high power density, and to be self reforming. A thin film electrolyte (1-2 microm thickness), e.g., yttria-stabilized zirconia (YSZ), is deposited on a nickel foil substrate. The electrolyte thin film is polycrystalline when deposited on a polycrystalline nickel foil substrate, and is (100) textured when deposited on an atomically textured nickel foil substrate. The Ni foil substrate is then converted into a porous SOFC anode by photolithographic patterning and etching to develop porosity. A composite La(0.5)Sr(0.5)CoO(3) cathode is then deposited on the thin film electrolyte. The resultant thin film hetero structure fuel cells have operated at a significantly reduced temperature: as low as 470 degrees C, with a maximum power density of 140 mW cm(-2) at 575 degrees C, and an efficiency of >50%. This drastic reduction in operating temperature for an SOFC now also allows for the use of hydrocarbon fuels without the need for a separate reformer as the nickel anode effectively dissociates hydrocarbons within this temperature range. These nanostructured fuel cells show excellent potential for high power density, small volume, high efficiency fuel cells for power generation applications.
Product Description
Nickel, foil, 25mm disks, thickness 0.30mm, as rolled, 99.98%
Nickel, foil, 2m coil, thickness 0.02mm, 99.9%
Nickel, foil, not light tested, 300x300mm, thickness 0.0125mm, 99.9%
Nickel, foil, not light tested, 148x300mm, thickness 0.0025mm, 99.95%
Nickel, foil, 150x150mm, thickness 0.15mm, annealed, 99%
Nickel, foil, not light tested, 150x150mm, thickness 0.0125mm, 99.9%
Nickel, foil, 25mm disks, thickness 0.5mm, annealed, 99.98%
Nickel, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.98%
Nickel, foil, not light tested, 25x25mm, thickness 0.00075mm, temporary acrylic support, 99.95%
Nickel, foil, not light tested, 300x300mm, thickness 0.0125mm, annealed, 99%
Nickel, foil, light tested, 50x50mm, thickness 0.075mm, as rolled, 99.98%
Nickel, foil, light tested, 50x50mm, thickness 0.0125mm, 99.9%
Nickel, foil, not light tested, 50x50mm, thickness 0.001mm, 99.95%
Nickel, foil, not light tested, 25x25mm, thickness 0.0025mm, 99.95%
Nickel, foil, light tested, 300x300mm, thickness 0.02mm, 99.9%
Nickel, foil, 25x25mm, thickness 0.3mm, annealed, 99%
Nickel, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99.9%
Nickel, foil, not light tested, 45x45mm, thickness 0.004mm, hard, 99.98%
Nickel, foil, not light tested, 100x100mm, thickness 0.0025mm, 99.95%
Nickel, foil, 100x100mm, thickness 0.3mm, annealed, 99%
Nickel, foil, 2m coil, thickness 0.05mm, 99.9%
Nickel, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Nickel, foil, light tested, 50x50mm, thickness 0.007mm, 99.95%
Nickel, foil, not light tested, 50x50mm, thickness 0.00025mm, temporary acrylic support, 99.95%
Nickel, foil, not light tested, 150x150mm, thickness 0.008mm, 99.95%
Nickel, foil, not light tested, 100x100mm, thickness 0.008mm, 99.95%
Nickel, foil, 2m coil, thickness 0.025mm, coil width 600mm, as rolled, 99%
Nickel, foil, 2m coil, thickness 0.0075mm, 99.95%
Nickel, foil, not light tested, 25x25mm, thickness 0.003mm, 99.95%
Nickel, foil, 2m coil, thickness 0.015mm, 99.9%