Merck
  • Lack of an effect of antibiotic treatment on prolonged detection of chlamydial DNA in murine genital tract infection.
Lack of an effect of antibiotic treatment on prolonged detection of chlamydial DNA in murine genital tract infection.
Antimicrobial agents and chemotherapy (2007-05-16)
Dawn M Reeves, Uma Nagarajan, Catherine O'Connell, Charles W Andrews, Toni Darville
ABSTRACT
Mice treated with antibiotics early or late after active infection had resolved were examined for chlamydial DNA in endocervical swabs. The early eradication of infection limited oviduct pathology, despite the continued detection of chlamydial DNA by nested PCR. Late antibiotic treatment had no effect on the ability to detect DNA or oviduct pathology.
Sigma-Aldrich
REDExtract-N-Amp Blood PCR Kit, sufficient for 10 extractions, sufficient for 10 amplifications
Sigma-Aldrich
REDExtract-N-Amp Blood PCR Kit, sufficient for 1000 extractions, sufficient for 1000 amplifications
Sigma-Aldrich
REDExtract-N-Amp Blood PCR Kit, sufficient for 100 extractions, sufficient for 100 amplifications
Sigma-Aldrich
REDExtract-N-Amp Blood PCR Kit, sufficient for 100 extractions, sufficient for 500 amplifications
Sigma-Aldrich
Neutralization Solution for Blood, sufficient for 100 reactions, sufficient for 1000 reactions, for molecular biology
Sigma-Aldrich
Lysis solution for blood, sufficient for 100 reactions, for molecular biology