Merck
  • [A laboratory method for sensitized isomerization of polyene macrolides].
[A laboratory method for sensitized isomerization of polyene macrolides].
Antibiotiki i khimioterapiia = Antibiotics and chemoterapy [sic] (1996-02-01)
Iu D Shenin, A I Filippova
ABSTRACT
The study was aimed at the development of a laboratory procedure for isomerization of polyene macrolides in the presence of sensitizers. The reaction was performed with exposure of levorin solutions to the xenon lamp irradiation in the presence of rose bengal B as a sensitizers. It was shown that levoristatin contain in the levorin sample was not degraded under the experimental conditions. The use of a redetermined variant of the Vierordt's methods made it possible to offer a design formula for the estimation of the isolevorin content in solutions in the presence of levorin.
Sigma-Aldrich
Rose Bengal lactone, 95%
Sigma-Aldrich
Rose bengal, Dye content 95 %, ≥96% (HPLC)
Sigma-Aldrich
Rose bengal, certified by the BSC, Dye content ≥80 %