Merck
  • Demethylation kinetics of aspartame and L-phenylalanine methyl ester in aqueous solution.
Demethylation kinetics of aspartame and L-phenylalanine methyl ester in aqueous solution.
Pharmaceutical research (1993-08-01)
R D Skwierczynski, K A Connors
ABSTRACT
The kinetics of demethylation of aspartame and L-phenylalanine methyl ester were studied in aqueous solution at 25 degrees C over the pH range 0.27-11.5. The pseudo-first-order rate constant for aspartame was resolved into individual contributions from methyl ester hydrolysis and diketopiperazine formation. pH-rate profiles were quantitatively described by chemically reasonable kinetic schemes. Aspartame is maximally stable at pH 4 (t90 = 53 days at 25 degrees C); phenylalanine methyl ester, at pH 3. The potentiometrically measured pKa values were pKa1 3.19 and pKa2 7.87 for aspartame and pKa 7.11 for phenylalanine methyl ester.
Sigma-Aldrich
L-Phenylalanine methyl ester hydrochloride
Sigma-Aldrich
L-Phenylalanine methyl ester hydrochloride, 98%
Sigma-Aldrich
D-Phenylalanine methyl ester hydrochloride, 98%