Merck
  • Percutaneous absorption of benzyl acetate through rat skin in vitro. 3. A comparison with human skin.
Percutaneous absorption of benzyl acetate through rat skin in vitro. 3. A comparison with human skin.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (1994-11-01)
A Garnett, S A Hotchkiss, J Caldwell
ABSTRACT
The comparative absorption of the fragrance and industrial compound, benzyl acetate, has been studied in rat and human skin, using shaved, full-thickness dorsal skin of male Fischer 344 rats and full-thickness human skin obtained from patients undergoing surgical resection. Penetration of the compound through rat and human skin was evaluated in vitro in flow-through diffusion cells following topical application of neat [methylene-14C] benzyl acetate (33.1 mg/cm2) to the epidermal surface and occlusion with a teflon cap, 2.9 cm above the skin surface. The absorption of benzyl acetate across rat skin was rapid and extensive, reaching 34.3 +/- 3.9% of the applied dose (11.3 +/- 1.3 mg/cm2) (mean +/- SD, n = 12) at 24 hr and 55.8 +/- 5.0% of the applied dose (18.5 +/- 1.7 mg/cm2) at 72 hr. The penetration of benzyl acetate was significantly (P < 0.05) less rapid and extensive through human skin, reaching 5.5 +/- 0.1% of the applied dose (1.8 +/- 0.0 mg/cm2) (mean +/- SD, n = 12) at 24 hr and 17.8 +/- 3.3% of the applied dose (5.9 +/- 1.1 mg/cm2) at 72 hr. The rate of penetration of benzyl acetate was greater through rat skin than through human tissue at all time points studied up to 72 hr. The maximum rate of skin penetration was 0.6 +/- 0.1 mg/cm2/hr and 0.1 +/- 0.0 mg/cm2/hr through rat and human skin, respectively. These data indicate that systemic exposure to benzyl acetate may occur after skin contact in humans. They also support the evidence from the literature that human skin is generally less permeable to xenobiotics than rat skin.
Product Description
Supelco
Benzyl acetate, analytical standard
Sigma-Aldrich
Benzyl acetate, ≥99%
Sigma-Aldrich
Benzyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Benzyl acetate, natural, ≥99%, FCC, FG