Merck
  • Disseminated intravascular coagulation (DIC) induced by liquoid (polyanetholsulfonate) in the rat. V. Effects on circulating fibronectin.
Disseminated intravascular coagulation (DIC) induced by liquoid (polyanetholsulfonate) in the rat. V. Effects on circulating fibronectin.
British journal of experimental pathology (1986-10-01)
J Cerda, R E Urizar, R Conran, F A Blumenstock, J E Kaplan, A B Malik, R Simon
ABSTRACT
Levels of plasma fibronectin (Fn) were 63% lower than normal 15 min after the intravenous injection of liquoid (P less than 0.01); 3 h later they were still low but rebounded to 35% above normal (P less than 0.01) by 24 h. Concurrently microthrombi containing fibrinogen, Fn and Factor VIII related-antigens (VIII:Ag) were detected in the kidneys and lungs by immunohistopathological studies. Ultrastructurally, thrombi were composed of dense granular and occasional fibrillar non-striated material. In liquoid-injected rats 125I-fibrinogen mainly localized in kidneys and lungs, especially in the latter (P less than 0.01), and the lungs had a higher wet-to-dry weight ratios than did controls (P less than 0.01). It is concluded that the polyanion (liquoid)-induced intravascular coagulation-like reaction sequestered Fn concomitantly with the precipitation of fibrinogen and VII:Ag in the microclots. The reduced concentration of plasma Fn may have impaired the disposal of coagulation products thus enhancing the expression of the coagulopathy-mediated renal and pulmonary histopathology. It is suggested that the liquoid-related coagulopathy may have resulted in enzymatic lysis of Fn.
Sigma-Aldrich
Polyanetholesulfonic acid sodium salt, powder
Sigma-Aldrich
Polyanetholesulfonic acid sodium salt, BioUltra
Sigma-Aldrich
Polyanetholesulfonic acid sodium salt