Merck
  • Thy1-expressing neurons in the basolateral amygdala may mediate fear inhibition.
Thy1-expressing neurons in the basolateral amygdala may mediate fear inhibition.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2013-06-21)
Aaron M Jasnow, David E Ehrlich, Dennis C Choi, Joanna Dabrowska, Mallory E Bowers, Kenneth M McCullough, Donald G Rainnie, Kerry J Ressler
ABSTRACT
Research has identified distinct neuronal circuits within the basolateral amygdala (BLA) that differentially mediate fear expression versus inhibition; however, molecular markers of these populations remain unknown. Here we examine whether optogenetic activation of a cellular subpopulation, which may correlate with the physiologically identified extinction neurons in the BLA, would differentially support fear conditioning versus fear inhibition/extinction. We first molecularly characterized Thy1-channelrhodopsin-2 (Thy1-ChR2-EYFP)-expressing neurons as a subpopulation of glutamatergic pyramidal neurons within the BLA. Optogenetic stimulation of these neurons inhibited a subpopulation of medial central amygdala neurons and shunted excitation from the lateral amygdala. Brief activation of these neurons during fear training disrupted later fear memory in male mice. Optogenetic activation during unreinforced stimulus exposure enhanced extinction retention, but had no effect on fear expression, locomotion, or open-field behavior. Together, these data suggest that the Thy1-expressing subpopulation of BLA pyramidal neurons provide an important molecular and pharmacological target for inhibiting fear and enhancing extinction and for furthering our understanding of the molecular mechanisms of fear processing.
Sigma-Aldrich
L-Glutamic acid hydrochloride
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, FCC
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland