Merck
  • Alternative mutational pathways to intermediate resistance to vancomycin in methicillin-resistant Staphylococcus aureus.
Alternative mutational pathways to intermediate resistance to vancomycin in methicillin-resistant Staphylococcus aureus.
The Journal of infectious diseases (2013-03-30)
Celine Vidaillac, Susana Gardete, Ryan Tewhey, George Sakoulas, Glenn W Kaatz, Warren E Rose, Alexander Tomasz, Michael J Rybak
ABSTRACT
We used 2 in vitro experimental systems to compare phenotypic and genotypic changes that accompany selection of mutants of methicillin-resistant Staphylococcus aureus (MRSA) strain JH1 with low-level vancomycin resistance similar to the type found in vancomycin-intermediate S. aureus (VISA). The previously described MRSA strain JH1 and its vancomycin-intermediate mutant derivative JH2, both of which were recovered from a patient undergoing vancomycin chemotherapy, were used in this study. Mutants of JH1 were selected in vitro by means of a pharmacokinetic/pharmacodynamic (PK/PD) model of simulated endocardial vegetations (SEVs) and by exposure to vancomycin in laboratory growth medium. Phenotypic abnormalities of JH1 mutants generated by each in vitro experimental system were compared to those of JH2, and whole genomes of 2 in vitro JH1 mutants were sequenced to identify mutations that may be associated with an increased vancomycin minimum inhibitory concentration. JH1R1 was selected from the PK/PD model, and JH1R2 was selected in laboratory growth medium. Both mutants displayed reduced vancomycin and daptomycin susceptibility and phenotypic alterations (eg, thicker cell walls and abnormal autolysis) that are typical of in vivo VISA mutants. Genome sequencing of JH1R1 identified point mutations in 4 genes, all of which were different from the mutations described in JH2, including 1 mutation in yycG, a component of the WalKR sensory regulatory system. Sequencing of the JH1R2 genome identified mutations in 7 genes, including 2 in rpoB. Our findings indicate that JH1 is able to develop VISA-type resistance through several alternative genetic pathways.
Product Description
Millipore
Vancomycin supplement, suitable for microbiology
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥900 μg per mg (as vancomycin base)
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, meets USP testing specifications
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Vancomycin hydrochloride from Streptomyces orientalis, ≥85% (Vancomycin B)