Merck
  • Palladium-catalyzed insertion of N-tosylhydrazones for the synthesis of isoindolines.
Palladium-catalyzed insertion of N-tosylhydrazones for the synthesis of isoindolines.
Chemical communications (Cambridge, England) (2013-03-14)
Ping-Xin Zhou, Jian-Yi Luo, Lian-Biao Zhao, Yu-Ying Ye, Yong-Min Liang
ABSTRACT
Isoindolines are synthesized by palladium-catalyzed coupling reaction of N-(2-iodobenzyl) anilines with α,β-unsaturated N-tosylhydrazones. The reaction has several potential advantages: (1) toleration of a wide range of functional groups, (2) easy to handle and with mild conditions, (3) enriches the isoindoline family, (4) two new bonds form in one step.
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Indoline, ReagentPlus®, 99%
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sigma-Aldrich
Aniline hydrochloride, ≥99%