Merck
  • Template-free bottom-up synthesis of yolk-shell vanadium oxide as high performance cathode for lithium ion batteries.
Template-free bottom-up synthesis of yolk-shell vanadium oxide as high performance cathode for lithium ion batteries.
Chemical communications (Cambridge, England) (2013-01-17)
Hongchang Pang, Peng Cheng, Hongbin Yang, Jinlin Lu, Chun Xian Guo, Guiling Ning, Chang Ming Li
ABSTRACT
A template-free strategy is exploited to bottom-up synthesize yolk-shell vanadium oxide through a two-step spontaneous assembly of hydrolytically formed subunits in a one-pot process. The unique structured vanadium pentoxide exhibits excellent cathode performance for lithium ion batteries.
Sigma-Aldrich
Vanadium(V) oxide, 99.9% trace metals basis
Sigma-Aldrich
Vanadium(V) oxide, ≥98%
Sigma-Aldrich
Vanadium(V) oxide, ≥99.6% trace metals basis