Merck
  • Binding of naproxen enantiomers to human serum albumin studied by fluorescence and room-temperature phosphorescence.
Binding of naproxen enantiomers to human serum albumin studied by fluorescence and room-temperature phosphorescence.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-01-09)
Ivonne Lammers, Virginie Lhiaubet-Vallet, Freek Ariese, Miguel A Miranda, Cees Gooijer
ABSTRACT
The interaction of the enantiomers of the non-steroidal anti-inflammatory drug naproxen (NPX) with human serum albumin (HSA) has been investigated using fluorescence and phosphorescence spectroscopy in the steady-state and time-resolved mode. The absorption, fluorescence excitation, and fluorescence emission spectra of (S)-NPX and (R)-NPX differ in shape in the presence of HSA, indicating that these enantiomers experience a different environment when bound. In solutions containing 0.2M KI, complexation with HSA results in a strongly increased NPX fluorescence intensity and a decreased NPX phosphorescence intensity due to the inhibition of the collisional interaction with the heavy atom iodide. Fluorescence intensity curves obtained upon selective excitation of NPX show 8-fold different slopes for bound and free NPX. No significant difference in the binding constants of (3.8±0.6)×10(5) M(-1) for (S)-NPX and (3.9±0.6)×10(5) M(-1) for (R)-NPX was found. Furthermore, the addition of NPX quenches the phosphorescence of the single tryptophan in HSA (Trp-214) based on Dexter energy transfer. The short-range nature of this mechanism explains the upward curvature of the Stern-Volmer plot observed for HSA: At low concentrations NPX binds to HSA at a distance from Trp-214 and no quenching occurs, whereas at high NPX concentrations the phosphorescence intensity decreases due to dynamic quenching by NPX diffusing into site I from the bulk solution. The dynamic quenching observed in the Stern-Volmer plots based on the longest phosphorescence lifetime indicates an overall binding constant to HSA of about 3×10(5) M(-1) for both enantiomers.
Sigma-Aldrich
Naproxen, meets USP testing specifications
Supelco
Naproxen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Naproxen, VETRANAL®, analytical standard
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
Naproxen, European Pharmacopoeia (EP) Reference Standard