Merck
  • Loss of the initial efficacy of levetiracetam in patients with refractory epilepsy.
Loss of the initial efficacy of levetiracetam in patients with refractory epilepsy.
Seizure (2013-01-03)
Gha-hyun Lee, Bo-mi Kim, Joong Koo Kang, Sang-Ahm Lee
ABSTRACT
The efficacy and safety of the anti-convulsive drug levetiracetam (LEV) has been well documented but few clinical studies have investigated tolerance to LEV. The aim of this study was to evaluate the loss of the initial efficacy of LEV in adult patients with refractory partial-onset seizures. We enrolled patients with refractory partial epilepsy who were started on add-on LEV treatment. The efficacy of LEV was evaluated every three months and the seizure frequency was decided by the average number of monthly seizures. A responder was defined as a patient with a ≥50% reduction in seizure frequency from the baseline. Seizure freedom was defined as a seizure-free status from the beginning of LEV treatment to the evaluation period. Loss of the initial efficacy was defined as a shift from responder status during the first three months of LEV treatment to non-responder status during the follow-up period. A total of 95 epilepsy patients were analyzed. During the first three months of LEV treatment, 50 (52.6%) of the 95 patients were responders with a ≥50% seizure reduction. Nine patients (18.0%) showed a loss of initial efficacy during the second three-month period. In contrast, only two (4.0%) of the non-responders during the first three months became responders during the next three months. However, this difference did not reach statistical significance (P=0.054). Based on Kaplan-Meier survival estimates, 49.2% of the patients who initially responded to LEV treatment during the first three months were predicted to lose this response at 42 months. Loss of the initial efficacy of LEV treatment occurred mostly within 18 months. This study suggests that the occurrence of tolerance is more common than late gain of efficacy of treatment although larger prospective studies would have to be carried out to prove this observation.
Sigma-Aldrich
Piracetam
Levetiracetam impurity D, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Levetiracetam, ≥98% (HPLC)
Supelco
Levetiracetam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Levetiracetam, European Pharmacopoeia (EP) Reference Standard